4-Acetoxybenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11350-18
A11350-30
A11350-14
CAAAAA11350-18EA
82.48
CAD
CAAAAA11350-18
CAAAAA11350-30
CAAAA11350-14
4-Acetoxybenzoic acid 98+%
4-Acetoxybenzoic acid
MDL: MFCD00002540
Beilstein Registry No.: 1871997
Purity: 98+%
CAS No.: 2345-34-8
Molecular Formula: C9H8O4
Formula Weight: 180.15
Melting Point: 192-195°
Beilstein Reference: 1871997
Formula:
C₉H₈O₄ Melting Pt: 192…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002540 CAS Number: 2345-34-8 EINECS: 219-066-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...