Butyrophenone 99%
Supplier: Thermo Scientific Chemicals
Warning
|
A11345-14
A11345-18
A11345-30
CAAAAA11345-14EA
55.93
CAD
CAAAAA11345-14
CAAAA11345-18
CAAAA11345-30
Butyrophenone 99%
Butyrophenone
MDL: MFCD00009397
Beilstein Registry No.: 508305
Purity: 99%
CAS No.: 495-40-9
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 88°(190°F)
Refractive Index: 1.5195
Odor: Characteristic
Melting Point: 11-13°
Boiling Point: 222°
Beilstein Reference: 508305
MDL No.: MFCD00009397
Formula:
C₆H₅C(O)(CH₂)₂CH₃ Boiling Pt: 222 °C Melting Pt: 11…13 °C Density: 1.021 Flash Pt: 88 °C(190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009397 CAS Number: 495-40-9 EINECS: 207-799-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...