3,4-Dimethoxyphenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11344-06
A11344-14
A11344-22
CAAAA11344-06EA
144.73
CAD
CAAAA11344-06
CAAAA11344-14
CAAAA11344-22
3,4-Dimethoxyphenol 98%
3,4-Dimethoxyphenol
MDL: MFCD00008390
Beilstein Registry No.: 1366650
Purity: 98%
CAS No.: 2033-89-8
Molecular Formula: C8H10O3
Formula Weight: 154.16
Melting Point: 79-82°
Boiling Point: 167°/14mm
MDL No.: MFCD00008390
Formula:
C₈H₁₀O₃ Boiling Pt: 167 °C/14 mm Melting Pt: 79…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008390 CAS Number: 2033-89-8 EINECS: 217-995-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...