1-Bromodecane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11340-22
A11340-0C
A11340-0E
A11340-36
CAAAAA11340-22EA
45.19
CAD
CAAAAA11340-22
CAAAA11340-0C
CAAAA11340-0E
CAAAA11340-36
1-Bromodecane 98%
1-Bromodecane
MDL: MFCD00000221
Beilstein Registry No.: 1735227
Purity: 98%
CAS No.: 112-29-8
Molecular Formula: C10H21Br
Formula Weight: 221.19
Flash Point: 94°(201°F)
Refractive Index: 1.4560
Melting Point: -30°
Boiling Point: 238°
Beilstein Reference: 1735227
UN No.: 2810
MDL No.: MFCD00000221
Formula:
BrH₂C(CH₂)₈CH₃ Boiling Pt: 238 °C Melting Pt: –30 °C Density: 1.067 Flash Pt: 94 °C (201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000221 CAS Number: 112-29-8 EINECS: 203-955-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...