1,2-Difluoro-4-iodobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11339-06
A11339-14
CAAAA11339-06EA
61.15
CAD
CAAAA11339-06
CAAAA11339-14
1,2-Difluoro-4-iodobenzene 99%
1,2-Difluoro-4-iodobenzene
MDL: MFCD00044630
Beilstein Registry No.: 2081099
Purity: 99%
CAS No.: 64248-58-4
Molecular Formula: C6H3F2I
Formula Weight: 239.99
Flash Point: 72°(161°F)
Refractive Index: 1.5590
Boiling Point: 177-178°
Beilstein Reference: 2081099
MDL No.: MFCD00044630
Formula:
C₆H₃F₂I Boiling Pt: 177…178 °C Density: 1,990 Flash Pt: 72 °C(161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044630 CAS Number: 64248-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...