1,3-Diethoxybenzene 95%

Supplier: Thermo Scientific Chemicals

A11338-09 A11338-18 A11338-30
CAAAA11338-09EA 52.61 CAD
CAAAA11338-09 CAAAA11338-18 CAAAA11338-30
1,3-Diethoxybenzene 95%
1,3-Diethoxybenzene

MDL: MFCD00015145 Beilstein Registry No.: 2517730


Purity: 95% CAS No.: 2049-73-2 Molecular Formula: C10H14O2 Formula Weight: 166.22 Refractive Index: 1.5095 Melting Point: 11-12° Boiling Point: 234-236° Beilstein Reference: 2517730 MDL No.: MFCD00015145

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 234…236 °C
Melting Pt: 11…12 °C
Density: 1.010
Storage Temperature: Ambient
MDL Number: MFCD00015145
CAS Number: 2049-73-2
EINECS: 218-071-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR