, (dried basis) 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
A11321-0I
A11321-0B
A11321-30
CAAAAA11321-0IEA
816.86
CAD
CAAAAA11321-0I
CAAAA11321-0B
CAAAA11321-30
, (dried basis) 98+%
di-Potassium hydrogen phosphate
MDL: MFCD00011383
Purity: 98+%
CAS No.: 7758-11-4
Molecular Formula: K2HPO4
Formula Weight: 174.18
Merck Reference: 14,7658
MDL No.: MFCD00011383
Formula:
K₂HPO₄ MW: 174.18 g/mol Melting Pt: 340 Storage Temperature: Ambient |
MDL Number:
MFCD00011383 CAS Number: 7758-11-4 EINECS: 231-834-5 Merck Index: 14,7658 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...