2,5-Dichloro-α,α,α-trifluorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,4-Dichloro-2-(trifluoromethyl)benzene, 2,5-Dichlorobenzotrifluoride
A11310-06
A11310-14
CAAAA11310-06EA
39.97
CAD
CAAAA11310-06
CAAAA11310-14
2,5-Dichloro-α,α,α-trifluorotoluene 98%
2,5-Dichloro-α,α,α-trifluorotoluene
MDL: MFCD00000608
Beilstein Registry No.: 1955442
Purity: 98%
CAS No.: 320-50-3
Molecular Formula: C7H3Cl2F3
Formula Weight: 215.00
Flash Point: 76°(168°F)
Refractive Index: 1.4830
Beilstein Reference: 1955442
UN No.: 1760
MDL No.: MFCD00000608
Formula:
C₇H₃Cl₂F₃ Boiling Pt: 181 °C (1013 hPa) Melting Pt: 11 °C Density: 1.483 Flash Pt: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000608 CAS Number: 320-50-3 EINECS: 206-276-0 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...