2,3-Dihydroxynaphthalene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11307-18
A11307-30
CAAAA11307-18EA
96.22
CAD
CAAAA11307-18
CAAAA11307-30
2,3-Dihydroxynaphthalene 98%
2,3-Dihydroxynaphthalene
MDL: MFCD00004073
Beilstein Registry No.: 742375
Purity: 98%
CAS No.: 92-44-4
Molecular Formula: C10H8O2
Formula Weight: 160.17
Flash Point: 175°(347°F)
Melting Point: 162-165°
Beilstein Reference: 742375
MDL No.: MFCD00004073
Formula:
C₁₀H₈O₂ Boiling Pt: 354 °C (1013 hPa) Melting Pt: 162…165 °C Density: 1.12 g/cm³ (20 °C) Flash Pt: 175 °C(347 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004073 CAS Number: 92-44-4 EINECS: 202-156-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...