4-Nitro-m-toluonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11301-09
A11301-06
A11301-14
CAAAAA11301-09EA
273.34
CAD
CAAAAA11301-09
CAAAA11301-03
CAAAA11301-06
CAAAA11301-14
4-Nitro-m-toluonitrile 97%
4-Nitro-m-toluonitrile
MDL: MFCD00017615
Beilstein Registry No.: 2443778
Purity: 97%
CAS No.: 96784-54-2
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Melting Point: 78-84°
Beilstein Reference: 2443778
UN No.: 3439
MDL No.: MFCD00017615
Formula:
C₈H₆N₂O₂ Melting Pt: 78…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017615 CAS Number: 96784-54-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...