2-Methyl-4-pentenoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11297-09
A11297-18
CAAAA11297-09EA
91.01
CAD
CAAAA11297-09
CAAAA11297-18
2-Methyl-4-pentenoic acid 98%
2-Methyl-4-pentenoic acid
MDL: MFCD00015570
Beilstein Registry No.: 1720987
Purity: 98%
CAS No.: 1575-74-2
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 107°(224°F)
Refractive Index: 1.4310
Odor: Characteristic
Boiling Point: 195°
Beilstein Reference: 1720987
UN No.: 3265
MDL No.: MFCD00015570
Formula:
C₆H₁₀O₂ Boiling Pt: 195 °C Density: 0.950 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015570 CAS Number: 1575-74-2 EINECS: 216-404-7 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...