Melamine 99%

Supplier: Thermo Scientific Chemicals

A11295-0C A11295-0E A11295-36
CAAAA11295-0CEA 369.72 CAD
CAAAA11295-0C CAAAA11295-0E CAAAA11295-36
Melamine 99%
Melamine

MDL: MFCD00006055 Beilstein Registry No.: 124341


Purity: 99% CAS No.: 108-78-1 Molecular Formula: C3H6N6 Formula Weight: 126.12 Melting Point: ca 350° dec. Beilstein Reference: 124341 Merck Reference: 14,5811 MDL No.: MFCD00006055

Formula: C₃H₆N₆
Melting Pt: ∼ 350 °C (dec.)
Density: 1.573
Flash Pt: ≥ 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00006055
CAS Number: 108-78-1
EINECS: 203-615-4
Merck Index: 14,5811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR