2,6-Difluorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11286-22
A11286-06
A11286-14
CAAAAA11286-22EA
349.18
CAD
CAAAAA11286-22
CAAAA11286-06
CAAAA11286-14
2,6-Difluorobenzoyl chloride 98%
2,6-Difluorobenzoyl chloride
MDL: MFCD00000659
Beilstein Registry No.: 639438
Purity: 98%
CAS No.: 18063-02-0
Molecular Formula: C7H3ClF2O
Formula Weight: 176.55
Flash Point: 72°(162°F)
Refractive Index: 1.4988
Boiling Point: 72-77°/13mm
Beilstein Reference: 639438
UN No.: 3265
MDL No.: MFCD00000659
Formula:
C₇H₃ClF₂O Boiling Pt: 72…77 °C/13 mm Density: 1.407 Flash Pt: 72 °C (162 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000659 CAS Number: 18063-02-0 EINECS: 241-971-2 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...