2-Acetyl-5-methylthiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11282-22
A11282-06
A11282-14
CAAAAA11282-22EA
1188.16
CAD
CAAAAA11282-22
CAAAA11282-06
CAAAA11282-14
2-Acetyl-5-methylthiophene 98%
2-Acetyl-5-methylthiophene
MDL: MFCD00014529
Beilstein Registry No.: 110854
Purity: 98%
CAS No.: 13679-74-8
Molecular Formula: C7H8OS
Formula Weight: 140.20
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5625
Melting Point: 24-28°
Boiling Point: 65-67°/1mm
Beilstein Reference: 110854
MDL No.: MFCD00014529
Formula:
C₇H₈OS Boiling Pt: 65…67 °C/1 mm Melting Pt: 24…28 °C Density: 1.119 Storage Temperature: Refrigerator |
MDL Number:
MFCD00014529 CAS Number: 13679-74-8 EINECS: 237-181-2 UN: 3335 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...