D-(-)-Tartaric acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
unnatural tartaric acid, (2S,3S)-2,3-Dihydroxysuccinic acid, (S,S)-(-)-Tartaric acid, (S,S)-(-)-2,3-Dihydroxysuccinic acid
A11264-36
A11264-14
A11264-22
CAAAAA11264-36EA
657.28
CAD
CAAAAA11264-36
CAAAA11264-14
CAAAA11264-22
D-(-)-Tartaric acid 99%
D(-)-Tartaric acid
Useful as a resolving agent
Purity: 99%
CAS No.: 147-71-7
Molecular Formula: C4H6O6
Formula Weight: 150.09
Flash Point: 210°(410°F)
Odor: Odorless
pH: 1.6
Melting Point: 166-170°
Beilstein Reference: 1725145
Merck Reference: 14,9068
Optical Rotation: -13° (c=10 in water)
MDL No.: MFCD00004238
Formula:
C₄H₆O₆ MW: 150.09 g/mol Boiling Pt: 356 °C (1013 hPa) Melting Pt: 166…170 °C Density: 1.76 g/cm³ (20 °C) Flash Pt: 210 °C(410 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004238 CAS Number: 147-71-7 EINECS: 205-695-6 Merck Index: 14,9068 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...