2-Bromo-4-fluorophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11263-06
A11263-14
A11263-22
CAAAA11263-06EA
76.16
CAD
CAAAA11263-06
CAAAA11263-14
CAAAA11263-22
2-Bromo-4-fluorophenol 98+%
2-Bromo-4-fluorophenol
MDL: MFCD00010614
Beilstein Registry No.: 2355593
Purity: 98+%
CAS No.: 496-69-5
Molecular Formula: C6H4BrFO
Formula Weight: 191.00
Flash Point: 85°(185°F)
Melting Point: 42-46°
Boiling Point: 123-125°/50mm
Beilstein Reference: 2355593
MDL No.: MFCD00010614
Formula:
C₆H₄BrFO Boiling Pt: 123…125 °C/50 mm Melting Pt: 42…46 °C Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010614 CAS Number: 496-69-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...