2-Phenoxynicotinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11247-03
A11247-06
A11247-14
CAAAAA11247-03EA
42.5
CAD
CAAAAA11247-03
CAAAA11247-06
CAAAA11247-14
2-Phenoxynicotinic acid 98%
2-Phenoxynicotinic acid
MDL: MFCD00014629
Beilstein Registry No.: 172906
Purity: 98%
CAS No.: 35620-71-4
Molecular Formula: C12H9NO3
Formula Weight: 215.21
Melting Point: 179-182°
Beilstein Reference: 172906
Formula:
C₁₂H₉NO₃ Melting Pt: 179…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014629 CAS Number: 35620-71-4 EINECS: 252-647-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...