Chroman-4-one 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Chromanone

A11242-14 A11242-22
CAAAA11242-14EA 320.74 CAD
CAAAA11242-14 CAAAA11242-22
Chroman-4-one 98+%
Chroman-4-one

MDL: MFCD00006840 Beilstein Registry No.: 124652


Purity: 98+% CAS No.: 491-37-2 Molecular Formula: C9H8O2 Formula Weight: 148.16 Flash Point: 110°(230°F) Refractive Index: 1.5750 Melting Point: 36-40° Boiling Point: 127-128°/13mm Beilstein Reference: 124652 MDL No.: MFCD00006840

Formula: C₉H₈O₂
Boiling Pt: 127…128 °C/13 mm
Melting Pt: 36…40 °C
Density: 1.1291 g/cm³ (99.8 °C)
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006840
CAS Number: 491-37-2
EINECS: 207-736-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR