Paracetamol 98%

Supplier: Thermo Scientific Chemicals
Warning

A11240-36 A11240-0B A11240-0I A11240-30
CAAAAA11240-36EA 122.92 CAD
CAAAAA11240-36 CAAAA11240-0B CAAAA11240-0I CAAAA11240-30
Paracetamol 98%
Paracetamol

Cyclooxygenase inhibitor and widely used analgesic


Purity: 98% CAS No.: 103-90-2 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 168-172° Beilstein Reference: 2208089 Merck Reference: 14,47 MDL No.: MFCD00002328

Formula: C₈H₉NO₂
Boiling Pt: 388 °C (1013 hPa)
Melting Pt: 168…172 °C
Density: 1.293
Storage Temperature: Ambient
MDL Number: MFCD00002328
CAS Number: 103-90-2
EINECS: 203-157-5
Merck Index: 14,47

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR