Methyl-2-chlorobenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Chlorobenzoic acid emthyl ester

A11237-14 A11237-22 A11237-36
CAAAA11237-14EA 62.09 CAD
CAAAA11237-14 CAAAA11237-22 CAAAA11237-36
Methyl-2-chlorobenzoate 98%
Methyl 2-chlorobenzoate

MDL: MFCD00016337 Beilstein Registry No.: 1364684


Purity: 98% CAS No.: 610-96-8 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Flash Point: 108°(226°F) Refractive Index: 1.5360 Boiling Point: 233-235° Beilstein Reference: 1364684 MDL No.: MFCD00016337

Formula: C₈H₇ClO₂
Boiling Pt: 233…235 °C
Density: 1.191
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016337
CAS Number: 610-96-8
EINECS: 210-242-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR