6-Aminonicotinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11223-06
A11223-14
CAAAA11223-03EA
90.94
CAD
CAAAA11223-03
CAAAA11223-06
CAAAA11223-14
6-Aminonicotinic acid 98%
6-Aminonicotinic acid
MDL: MFCD00006326
Beilstein Registry No.: 115992
Purity: 98%
CAS No.: 3167-49-5
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Melting Point: ca 320° dec.
Beilstein Reference: 115992
Merck Reference: 14,456
MDL No.: MFCD00006326
Formula:
C₆H₆N₂O₂ Melting Pt: ∼ 320 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006326 CAS Number: 3167-49-5 EINECS: 221-630-4 Merck Index: 14,456 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...