5-Bromo-1-pentene 96%

Supplier: Thermo Scientific Chemicals
Warning

A11219-06 A11219-14
CAAAAA11219-03EA 51.54 CAD
CAAAAA11219-03 CAAAA11219-06 CAAAA11219-14
5-Bromo-1-pentene 96%
5-Bromo-1-pentene
Formula: C₅H₉Br
Boiling Pt: 126…127 °C
Density: 1.259
Flash Pt: 30 °C (86 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000264
CAS Number: 1119-51-3
EINECS: 214-281-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR