2-Methyl-1,3-cyclohexanedione 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methylcyclohexane-1,3-dione
A11218-22
A11218-06
A11218-14
CAAAAA11218-22EA
579.86
CAD
CAAAAA11218-22
CAAAA11218-06
CAAAA11218-14
2-Methyl-1,3-cyclohexanedione 98+%
2-Methyl-1,3-cyclohexanedione
MDL: MFCD00001587
Beilstein Registry No.: 1281598
Purity: 98+%
CAS No.: 1193-55-1
Molecular Formula: C7H10O2
Formula Weight: 126.16
Storage Temperature: 2°C to 8°C
Melting Point: 204-208°
Beilstein Reference: 1281598
MDL No.: MFCD00001587
Formula:
C₇H₁₀O₂ Boiling Pt: 100 - 102 °C (8 -rr) Melting Pt: 204…208 °C Density: 1.313 g/cm³ (-150.15 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001587 CAS Number: 1193-55-1 EINECS: 214-773-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...