(3-Chlorophenyl)acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11210-06 A11210-14 A11210-22
CAAAA11210-06EA 48.35 CAD
CAAAA11210-06 CAAAA11210-14 CAAAA11210-22
(3-Chlorophenyl)acetic acid 98+%
(3-Chlorophenyl)acetic acid

MDL: MFCD00004332 Beilstein Registry No.: 2045103


Purity: 98+% CAS No.: 1878-65-5 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Odor: Odorless Melting Point: 77-80° Beilstein Reference: 2045103 MDL No.: MFCD00004332

Formula: (C₆H₄Cl)CH₂COOH
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00004332
CAS Number: 1878-65-5
EINECS: 217-520-0
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR