Ammonium iron (III) citrate, red brown

Supplier: Thermo Scientific Chemicals

A11199-0B A11199-0I A11199-30
CAAAA11199-0BEA 130.35 CAD
CAAAA11199-0B CAAAA11199-0I CAAAA11199-30
Ammonium iron (III) citrate, red brown
Ammonium iron (III) citrate

MDL: MFCD00013099


CAS No.: 1185-57-5 Odor: Weak Merck Reference: 14,4017 MDL No.: MFCD00013099

Formula: C₆H(₅+₄y)Fe(x)N(y)O₇
Boiling Pt: 197 °C (1013 hPa)
Density: 1.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6
Merck Index: 14,4017

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR