3,5-Dimethylbenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11198-09 A11198-30
CAAAA11198-09EA 59.88 CAD
CAAAA11198-09 CAAAA11198-30
3,5-Dimethylbenzoic acid 98+%
3,5-Dimethylbenzoic acid

MDL: MFCD00002525 Beilstein Registry No.: 1072182


Purity: 98+% CAS No.: 499-06-9 Molecular Formula: C9H10O2 Formula Weight: 150.18 Odor: Odorless Melting Point: 168-172° Beilstein Reference: 1072182 MDL No.: MFCD00002525

Formula: C₉H₁₀O₂
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 168…172 °C
Density: 1.211 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002525
CAS Number: 499-06-9
EINECS: 207-876-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR