2-Aminonicotinic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11195-06
A11195-14
A11195-22
CAAAA11195-06EA
75.05
CAD
CAAAA11195-06
CAAAA11195-14
CAAAA11195-22
2-Aminonicotinic acid 98+%
2-Aminonicotinic acid
MDL: MFCD00006318
Beilstein Registry No.: 119031
Purity: 98+%
CAS No.: 5345-47-1
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Melting Point: ca 298° dec.
Beilstein Reference: 119031
Formula:
C₆H₆N₂O₂ Melting Pt: ∼ 298 °C (dec.) Density: 1.545 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006318 CAS Number: 5345-47-1 EINECS: 226-296-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...