4-Nitrophenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Isocyanato-4-nitrobenzene

A11194-06 A11194-14
CAAAA11194-06EA 65.41 CAD
CAAAA11194-06 CAAAA11194-14
4-Nitrophenyl isocyanate 97%
4-Nitrophenyl isocyanate

MDL: MFCD00007306 Beilstein Registry No.: 389516


Purity: 97% CAS No.: 100-28-7 Molecular Formula: C7H4N2O3 Formula Weight: 164.12 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 56-59° Boiling Point: 137-138°/11mm Beilstein Reference: 389516 UN No.: 3335 MDL No.: MFCD00007306

Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
Boiling Pt: 277
Melting Pt: 56…59 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007306
CAS Number: 100-28-7
EINECS: 202-836-3
UN: 3335

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR