2-Mercapto-5-methylbenzimidazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11192-06
A11192-14
A11192-22
CAAAAA11192-06EA
39.03
CAD
CAAAAA11192-06
CAAAA11192-14
CAAAA11192-22
2-Mercapto-5-methylbenzimidazole 98%
2-Mercapto-5-methylbenzimidazole
MDL: MFCD00010617
Beilstein Registry No.: 5083
Purity: 98%
CAS No.: 27231-36-3
Molecular Formula: C8H8N2S
Formula Weight: 164.23
Melting Point: ca 295° dec.
Beilstein Reference: 5083
Formula:
C₈H₈N₂S Boiling Pt: 349 °C (1013 hPa) Melting Pt: ∼ 295 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010617 CAS Number: 27231-36-3 EINECS: 248-350-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...