4-Fluorobenzophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

A11185-14 A11185-22 A11185-36
CAAAA11185-14EA 79.32 CAD
CAAAA11185-14 CAAAA11185-22 CAAAA11185-36
4-Fluorobenzophenone 97%
4-Fluorobenzophenone

MDL: MFCD00000352 Beilstein Registry No.: 1869800


Purity: 97% CAS No.: 345-83-5 Molecular Formula: C13H9FO Formula Weight: 200.21 Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 130-132°/3mm Beilstein Reference: 1869800 MDL No.: MFCD00000352

Formula: C₁₃H₉FO
MW: 200.21 g/mol
Boiling Pt: 130…132 °C/3 mm
Melting Pt: 45…49 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000352
CAS Number: 345-83-5
EINECS: 206-463-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR