Sodium dodecyl sulfate (SDS), (max. 1,5% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

A11183-14 A11183-22 A11183-36
CAAAA11183-14EA 76 CAD
CAAAA11183-14 CAAAA11183-22 CAAAA11183-36
Sodium dodecyl sulfate (SDS), (max. 1,5% H₂O) 99% (dry weight)
Sodium dodecyl sulphate (SDS)

Denatures and solubilizes proteins for electrophoresis


Purity: 99% CAS No.: 151-21-3 Molecular Formula: CH3(CH2)11OSO3Na Formula Weight: 288.38 Physical Form: Powder Appearance: White Melting Point: ca 206° Beilstein Reference: 3599286 Merck Reference: 14,8636 UN No.: 2926 MDL No.: MFCD00036175


Denatures and solubilizes proteins for electrophoresis

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: ∼ 206 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,8636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR