L(+)-Serine 99%
Supplier: Thermo Scientific Chemicals
A11179-14
A11179-22
A11179-36
CAAAA11179-14EA
66.41
CAD
CAAAA11179-14
CAAAA11179-22
CAAAA11179-36
L(+)-Serine 99%
L(+)-Serine
MDL: MFCD00064224
Beilstein Registry No.: 1721404
Fieser: 12,430 14,282
Optical Rotation: +14.5°C (c=9 in 1N HCl)
Purity: 99%
CAS No.: 56-45-1
Molecular Formula: C3H7NO3
Formula Weight: 105.09
Melting Point: 214-224° dec.
Beilstein Reference: 1721404
Merck Reference: 14,8460
Optical Rotation: +15° (c=9 in 1N HCl)
MDL No.: MFCD00064224
Formula:
C₃H₇NO₃ Boiling Pt: 259 °C (1013 hPa) Melting Pt: 214…224 °C (dec.) Density: 1.6 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064224 CAS Number: 56-45-1 EINECS: 200-274-3 Merck Index: 14,8460 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...