4-Penten-1-ol 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11178-22 A11178-06 A11178-14
CAAAAA11178-22EA 1050.7 CAD
CAAAAA11178-22 CAAAA11178-06 CAAAA11178-14
4-Penten-1-ol 98+%
4-Penten-1-ol

MDL: MFCD00002975 Beilstein Registry No.: 1560163

Formula: C₅H₁₀O
Boiling Pt: 134…137 °C
Density: 0.840
Flash Pt: 43 °C(109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002975
CAS Number: 821-09-0
EINECS: 212-473-2
UN: 1987
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR