Benzonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11173-22
A11173-0E
A11173-36
CAAAAA11173-22EA
58.3
CAD
CAAAAA11173-22
CAAAA11173-0E
CAAAA11173-36
Benzonitrile 99%
Benzonitrile
MDL: MFCD00001770
Beilstein Registry No.: 506893
Purity: 99%
CAS No.: 100-47-0
Molecular Formula: C7H5N
Formula Weight: 103.12
Flash Point: 71°(159°F)
Refractive Index: 1.5285
Melting Point: -13°
Boiling Point: 191-193°
Beilstein Reference: 506893
Merck Reference: 14,1097
UN No.: 2224
MDL No.: MFCD00001770
Formula:
C₇H₅N Boiling Pt: 191…193 °C Melting Pt: –13 °C Density: 1.007 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001770 CAS Number: 100-47-0 EINECS: 202-855-7 UN: 2224 ADR: 6.1,II Merck Index: 14,1097 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...