1-Decyne 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11171-06
A11171-14
A11171-22
CAAAA11171-06EA
48.51
CAD
CAAAA11171-06
CAAAA11171-14
CAAAA11171-22
1-Decyne 98%
1-Decyne
MDL: MFCD00009576
Beilstein Registry No.: 1236372
Purity: 98%
CAS No.: 764-93-2
Molecular Formula: HC≡C(CH2)7CH3
Formula Weight: 138.26
Flash Point: 50°(122°F)
Refractive Index: 1.4270
Odor: Petrol-like
Melting Point: -44°
Boiling Point: 174-176°
Beilstein Reference: 1236372
UN No.: 3295
MDL No.: MFCD00009576
Formula:
C₁₀H₁₈ MW: 138.25 g/mol Boiling Pt: 174…176 °C Melting Pt: –44 °C Density: 0.767 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009576 CAS Number: 764-93-2 EINECS: 212-132-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...