Vanillin 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Hydroxy-3-methoxybenzaldehyde
A11169-0E
A11169-22
A11169-36
CAAAA11169-0EEA
627.26
CAD
CAAAA11169-0E
CAAAA11169-22
CAAAA11169-36
Vanillin 99%
Vanillin
MDL: MFCD00006942
Beilstein Registry No.: 472792
Purity: 99%
CAS No.: 121-33-5
Molecular Formula: C8H8O3
Formula Weight: 152.15
Flash Point: 153°(307°F)
Physical Form: Crystalline powder
Density: 1.06
Sensitivity: Air and Light Sensitive
Appearance: White
Melting Point: 81-83°
Boiling Point: 170°/15mm
Beilstein Reference: 472792
Merck Reference: 14,9932
MDL No.: MFCD00006942
Formula:
C₈H₈O₃ Boiling Pt: 170 °C/15 mm Melting Pt: 81…83 °C Density: 1.06 Flash Pt: 153 °C(307 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006942 CAS Number: 121-33-5 EINECS: 204-465-2 Merck Index: 14,9932 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...