4'-Methoxyacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

A11162-14 A11162-0B A11162-30
CAAAAA11162-14EA 37.92 CAD
CAAAAA11162-14 CAAAA11162-0B CAAAA11162-30
4'-Methoxyacetophenone 99%
4'-Methoxyacetophenone

MDL: MFCD00008745 Beilstein Registry No.: 742313


Purity: 99% CAS No.: 100-06-1 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 138°(280°F) Odor: Characteristic Boiling Point: 258-260° Beilstein Reference: 742313 MDL No.: MFCD00008745

Formula: C₉H₁₀O₂
Boiling Pt: 258…260 °C
Melting Pt: 36…40 °C
Density: 1.08
Flash Pt: 138 °C(280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008745
CAS Number: 100-06-1
EINECS: 202-815-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR