3-Iodophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A11155-09 A11155-18
CAAAA11155-09EA 82.63 CAD
CAAAA11155-09 CAAAA11155-18 CAAAA11155-30
3-Iodophenol 98%
3-Iodophenol

MDL: MFCD00002261 Beilstein Registry No.: 2039304 Fieser: 11,270


Purity: 98% CAS No.: 626-02-8 Molecular Formula: C6H5IO Formula Weight: 220.01 Flash Point: 110°(230°F) Melting Point: 38-43° Beilstein Reference: 2039304 MDL No.: MFCD00002261

Formula: C₆H₅IO
MW: 220.01 g/mol
Melting Pt: 38…43 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002261
CAS Number: 626-02-8
EINECS: 210-923-2
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR