2,5-Dichloroaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

A11151-0E A11151-36
CAAAA11151-0EEA 488.22 CAD
CAAAA11151-0E CAAAA11151-36
2,5-Dichloroaniline 99%
2,5-Dichloroaniline

MDL: MFCD00007667 Beilstein Registry No.: 1447438


Purity: 99% CAS No.: 95-82-9 Molecular Formula: C6H5Cl2N Formula Weight: 162.02 Flash Point: 139°(282°F) Melting Point: 48-52° Boiling Point: 250-251° Beilstein Reference: 1447438 UN No.: 3442 MDL No.: MFCD00007667

Formula: C₆H₅Cl₂N
Boiling Pt: 250…251 °C
Melting Pt: 48…52 °C
Density: 1.54
Flash Pt: 139 °C (282 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007667
CAS Number: 95-82-9
EINECS: 202-455-2
UN: 3442
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR