1-Octene 97+%

Supplier: Thermo Scientific Chemicals
Danger

A11146-0F A11146-AP
CAAAA11146-0FEA 164.32 CAD
CAAAA11146-0F CAAAA11146-AP
1-Octene 97+%
1-Octene

MDL: MFCD00009548 Beilstein Registry No.: 1734497


Purity: 97+% CAS No.: 111-66-0 Molecular Formula: C8H16 Formula Weight: 112.22 Flash Point: 21°(70°F) Refractive Index: 1.4090 Odor: Petroleum-like Melting Point: -102° Boiling Point: 122-123° Beilstein Reference: 1734497 Merck Reference: 14,1764 UN No.: 3295 MDL No.: MFCD00009548

Formula: C₈H₁₆
Boiling Pt: 122…123 °C
Melting Pt: –102 °C
Density: 0.715
Flash Pt: 21 °C (70 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009548
CAS Number: 111-66-0
EINECS: 203-893-7
UN: 3295
ADR: 3,III
Merck Index: 14,1764

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR