α-Bromo-3-iodotoulene 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11140-06
A11140-14
A11140-22
CAAAA11140-06EA
149.15
CAD
CAAAA11140-06
CAAAA11140-14
CAAAA11140-22
α-Bromo-3-iodotoulene 96%
MDL: MFCD00019016
Beilstein Registry No.: 742268
Purity: 96%
CAS No.: 49617-83-6
Molecular Formula: C7H6BrI
Formula Weight: 296.94
Flash Point:
Formula:
C₇H₆BrI Boiling Pt: 106 °C/1 mm Melting Pt: 48…51 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019016 CAS Number: 49617-83-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...