1-Bromo-2,2-dimethoxypropane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11139-09
A11139-18
A11139-30
CAAAA11139-09EA
90.22
CAD
CAAAA11139-09
CAAAA11139-18
CAAAA11139-30
1-Bromo-2,2-dimethoxypropane 98%
1-Bromo-2,2-dimethoxypropane
MDL: MFCD00000208
Beilstein Registry No.: 2036095
Purity: 98%
CAS No.: 126-38-5
Molecular Formula: C5H11BrO2
Formula Weight: 183.05
Flash Point: 40°(104°F)
Refractive Index: 1.4470
Boiling Point: 83-87°/80mm
Beilstein Reference: 2036095
UN No.: 3271
MDL No.: MFCD00000208
Formula:
BrH₂CC(OCH₃)₂CH₃ Boiling Pt: 83…87 °C/80 mm Density: 1.354 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000208 CAS Number: 126-38-5 EINECS: 204-784-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...