Dopamine hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(2-Aminoethyl)-3,4-dihydroxybenzene HCl, 3-Hydroxytyraminium chloride, 3-Hydroxytyramine hydrochloride, 3,4-Dihydroxyphenethylammonium chloride

A11136-06 A11136-14 A11136-22
CAAAA11136-06EA 68.26 CAD
CAAAA11136-06 CAAAA11136-14 CAAAA11136-22
Dopamine hydrochloride 99%
Dopamine hydrochloride

A neurotransmitter and vasopressor that modulates cortical activation


Purity: 99% CAS No.: 62-31-7 Molecular Formula: C8H11NO2.HCl Formula Weight: 189.64 Storage Temperature: 2°C to 8°C Melting Point: 243-250° Beilstein Reference: 3656720 Merck Reference: 14,3421 MDL No.: MFCD00012898


A neurotransmitter and vasopressor that modulates cortical activation

Formula: C₈H₁₂ClNO₂
MW: 189.64
Melting Pt: 243…250 °C
Density: 1.356
Storage Temperature: Refrigerator
MDL Number: MFCD00012898
CAS Number: 62-31-7
EINECS: 200-527-8
Merck Index: 14,3421

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR