Phthalimide potassium salt 98+%

Supplier: Thermo Scientific Chemicals

A11134-0E A11134-22 A11134-36
CAAAAA11134-0EEA 657.28 CAD
CAAAAA11134-0E CAAAA11134-22 CAAAA11134-36
Phthalimide potassium salt 98+%
Phthalimide potassium salt

MDL: MFCD00005887 Beilstein Registry No.: 3598719


Purity: 98+% CAS No.: 1074-82-4 Molecular Formula: C8H4KNO2 Formula Weight: 185.23 Melting Point: 300° Beilstein Reference: 3598719

Formula: C₈H₄KNO₂
Melting Pt: >300 °C
Density: 1.63
Storage Temperature: Ambient
MDL Number: MFCD00005887
CAS Number: 1074-82-4
EINECS: 214-046-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR