1-Heptyne 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11130-14
A11130-22
CAAAA11130-14EA
110.92
CAD
CAAAA11130-14
CAAAA11130-22
1-Heptyne 99%
1-Heptyne
MDL: MFCD00009529
Beilstein Registry No.: 1733518
Purity: 99%
CAS No.: 628-71-7
Molecular Formula: CH3(CH2)4C≡CH
Formula Weight: 96.17
Flash Point: -10°(14°F)
Refractive Index: 1.4095
Odor: Unpleasant
Melting Point: -81°
Boiling Point: 98-100°
Beilstein Reference: 1733518
UN No.: 3295
MDL No.: MFCD00009529
Formula:
C₇H₁₂ Boiling Pt: 98…100 °C Melting Pt: –81 °C Density: 0.736 Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009529 CAS Number: 628-71-7 EINECS: 211-051-5 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...