4-Hydroxy-2-methoxybenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11127-03
A11127-06
A11127-14
CAAAA11127-03EA
98.91
CAD
CAAAA11127-03
CAAAA11127-06
CAAAA11127-14
4-Hydroxy-2-methoxybenzaldehyde 98%
4-Hydroxy-2-methoxybenzaldehyde
MDL: MFCD00051964
Beilstein Registry No.: 2045122
Purity: 98%
CAS No.: 18278-34-7
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 156-160°
Beilstein Reference: 2045122
Formula:
C₈H₈O₃ MW: 152.15 Boiling Pt: 273 Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051964 CAS Number: 18278-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...