Cyclopropanecarbonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11125-09
A11125-18
A11125-30
CAAAA11125-09EA
79.79
CAD
CAAAA11125-09
CAAAA11125-18
CAAAA11125-30
Cyclopropanecarbonitrile 98%
Cyclopropanecarbonitrile
MDL: MFCD00001269
Beilstein Registry No.: 635790
Purity: 98%
CAS No.: 5500-21-0
Molecular Formula: C4H5N
Formula Weight: 67.09
Flash Point: 32°(89°F)
Refractive Index: 1.4210
Odor: Recognizable
Boiling Point: 134-135°
Beilstein Reference: 635790
UN No.: 3275
MDL No.: MFCD00001269
Formula:
C₄H₅N Boiling Pt: 134…135 °C Melting Pt: –25 °C Density: 0.905 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001269 CAS Number: 5500-21-0 EINECS: 226-836-8 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...