2,2'-Dipyridyl disulfide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Di(pyridin-2-yl)disulphane, 1,2-Di(pyridin-2-yl)disulfane, 2,2'-Dithiodipyridine

A11118-06 A11118-14 A11118-22
CAAAA11118-06EA 122.29 CAD
CAAAA11118-06 CAAAA11118-14 CAAAA11118-22
2,2'-Dipyridyl disulfide 98%
2,2'-Dipyridyl disulphide

MDL: MFCD00006287 Beilstein Registry No.: 154629 Fieser: 5,285 6,246


Purity: 98% CAS No.: 2127-03-9 Molecular Formula: C10H8N2S2 Formula Weight: 220.32 Storage Temperature: 2°C to 8°C Flash Point: 210°(410°F) Melting Point: 56-59° Beilstein Reference: 154629 MDL No.: MFCD00006287

Formula: C₁₀H₈N₂S₂
MW: 220.32 g/mol
Boiling Pt: 348
Melting Pt: 56…59 °C
Flash Pt: 210 °C (410 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006287
CAS Number: 2127-03-9
EINECS: 218-343-1
UN: 3335

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR