tert-Butyl acetoacetate 98+%
Supplier: Thermo Scientific Chemicals
Warning
|
A11115-22
A11115-36
CAAAA11115-22EA
67.47
CAD
CAAAA11115-22
CAAAA11115-36
tert-Butyl acetoacetate 98+%
tert-Butyl acetoacetate
MDL: MFCD00008811
Beilstein Registry No.: 1680303
Fieser: 1,83
Purity: 98+%
CAS No.: 1694-31-1
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 76°(169°F)
Refractive Index: 1.4200
Odor: Pleasant
pH: 5.7
Melting Point: -38°
Boiling Point: 71-72°/11mm
Beilstein Reference: 1680303
MDL No.: MFCD00008811
Formula:
C₈H₁₄O₃ Boiling Pt: 71…72 °C/11 mm Melting Pt: –38 °C Density: 0.961 Flash Pt: 76 °C(169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008811 CAS Number: 1694-31-1 EINECS: 216-904-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...