Diphenyl sulfide 98%

Supplier: Thermo Scientific Chemicals
Warning

A11114-14 A11114-22
CAAAA11114-14EA 59.41 CAD
CAAAA11114-14 CAAAA11114-22
Diphenyl sulfide 98%
Diphenyl sulphide

MDL: MFCD00003064 Beilstein Registry No.: 1907932 Fieser: 6,241


Purity: 98% CAS No.: 139-66-2 Molecular Formula: C12H10S Formula Weight: 186.28 Flash Point: 110°(230°F) Refractive Index: 1.6325 Odor: Unpleasant Melting Point: -40° Boiling Point: 295-296° Beilstein Reference: 1907932 Merck Reference: 14,7314 MDL No.: MFCD00003064

Formula: C₁₂H₁₀S
Boiling Pt: 295…296 °C
Melting Pt: –40 °C
Density: 1.118
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003064
CAS Number: 139-66-2
EINECS: 205-371-4
UN: 2810
ADR: 6.1,III
Merck Index: 14,7314

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR